Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myo6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004111.2 | NP_001004111.2 | 33 - 34 | 3672 | 67 | |
XM_005160309.4 | XP_005160366.1 | 33 - 34 | 3788 | 67 | ||
XM_005160311.4 | XP_005160368.1 | 31 - 32 | 3602 | 67 | ||
XM_009294500.3 | XP_009292775.1 | 34 - 35 | 3797 | 67 | ||
XM_009294501.3 | XP_009292776.1 | 33 - 34 | 3758 | 67 | ||
XM_009294502.3 | XP_009292777.1 | 32 - 33 | 3722 | 67 | ||
XM_021468404.1 | XP_021324079.1 | 34 - 35 | 3818 | 67 | ||
GenBank mRNA | AY618702.1 | - | 33 - 34 | 3670 | 67 | |
AY691329.1 | - | 31 - 32 | 3608 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Danio rerio Probe spans exons |