Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stmn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005923.2 | NP_001005923.2 | 2 - 3 | 268 | 76 | |
GenBank mRNA | BC083539.1 | - | 2 - 3 | 201 | 76 | |
BC164868.1 | - | 2 - 3 | 150 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |