Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrto4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017657.1 | NP_001017657.1 | 2 - 3 | 139 | 132 | |
GenBank mRNA | BC092913.1 | - | 2 - 3 | 139 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |