Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: txndc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020824.1 | NP_001018660.1 | 3 - 4 | 248 | 83 | |
GenBank mRNA | BC095367.1 | - | 3 - 4 | 248 | 83 | |
DQ285661.1 | - | 3 - 4 | 260 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |