Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtpbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024811.1 | NP_001019982.1 | 8 - 9 | 1540 | 65 | |
XM_005161694.4 | XP_005161751.1 | 9 - 10 | 1595 | 65 | ||
XM_009296201.3 | XP_009294476.1 | 9 - 10 | 1555 | 65 | ||
GenBank mRNA | BC056806.1 | - | 8 - 9 | 1538 | 65 | |
BC096803.1 | - | 8 - 9 | 1540 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |