Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025452.1 | NP_001020623.1 | 3 - 4 | 456 | 65 | |
GenBank mRNA | BC096804.1 | - | 3 - 4 | 456 | 65 | |
BC109403.1 | - | 3 - 4 | 402 | 65 | ||
BC165426.1 | - | 3 - 4 | 291 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |