Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044933.2 | NP_001038398.2 | 11 - 12 | 1265 | 103 | |
XM_005160726.4 | XP_005160783.1 | 11 - 12 | 1308 | 103 | ||
GenBank mRNA | BC129209.1 | - | 11 - 12 | 1265 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |