Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pard6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102675.1 | NP_001096145.1 | 2 - 3 | 286 | 131 | |
GenBank mRNA | EF550991.1 | - | 2 - 3 | 286 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Danio rerio Probe spans exons |