Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: faah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109825.1 | NP_001103295.1 | 14 - 15 | 1635 | 84 | |
GenBank mRNA | DQ986317.1 | - | 14 - 15 | 1635 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |