Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152949.1 | NP_694481.1 | 4 - 5 | 805 | 80 | |
XM_021480961.1 | XP_021336636.1 | 3 - 4 | 913 | 80 | ||
GenBank mRNA | AF234784.2 | - | 4 - 5 | 805 | 80 | |
BC045840.1 | - | 4 - 5 | 805 | 80 | ||
BC151890.1 | - | 4 - 5 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |