Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173260.1 | NP_775367.1 | 11 - 12 | 1107 | 85 | |
XM_005173300.4 | XP_005173357.1 | 11 - 12 | 85 | |||
GenBank mRNA | AF506232.1 | - | 11 - 12 | 1107 | 85 | |
BC071324.1 | - | 11 - 12 | 1071 | 85 | ||
BC154771.1 | - | 11 - 12 | 1241 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |