Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spaw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180967.2 | NP_851298.1 | 2 - 3 | 1066 | 123 | |
GenBank mRNA | AY240027.1 | - | 2 - 3 | 1066 | 123 | |
BC163609.1 | - | 2 - 3 | 1075 | 123 | ||
BC163616.1 | - | 2 - 3 | 1075 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |