Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200426.1 | NP_956720.1 | 7 - 8 | 918 | 80 | |
XM_017352203.2 | XP_017207692.1 | 6 - 7 | 958 | 80 | ||
XM_017352204.2 | XP_017207693.1 | 7 - 8 | 1381 | 80 | ||
GenBank mRNA | BC054668.1 | - | 7 - 8 | 918 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |