Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mxd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201056.1 | NP_957350.1 | 3 - 4 | 228 | 66 | |
GenBank mRNA | BC055579.1 | - | 3 - 4 | 228 | 66 | |
BC165925.1 | - | 3 - 4 | 239 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |