Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nucb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201493.1 | NP_958901.1 | 8 - 9 | 932 | 97 | |
GenBank mRNA | BC046077.1 | - | 8 - 9 | 932 | 97 | |
BC065437.1 | - | 8 - 9 | 898 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |