Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213375.1 | NP_998540.1 | 1 - 2 | 499 | 90 | |
XM_005157511.4 | XP_005157568.1 | 2 - 3 | 432 | 90 | ||
GenBank mRNA | BC046056.1 | - | 1 - 2 | 499 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |