Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kirrel3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099344.2 | NP_001092814.2 | 9 - 10 | 1790 | 61 | |
XM_021481418.1 | XP_021337093.1 | 9 - 10 | 1826 | 61 | ||
GenBank mRNA | CU638818.1 | - | 9 - 10 | 1203 | 61 | |
EF571006.1 | - | 9 - 10 | 1790 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |