Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lsm14aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200425.2 | NP_956719.2 | 6 - 7 | 991 | 75 | |
XM_005163175.2 | XP_005163232.1 | 5 - 6 | 972 | 75 | ||
XM_021470461.1 | XP_021326136.1 | 7 - 8 | 1150 | 75 | ||
XM_021470462.1 | XP_021326137.1 | 6 - 7 | 1056 | 75 | ||
GenBank mRNA | BC065337.1 | - | 6 - 7 | 991 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |