Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fshb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205624.1 | NP_991187.1 | 1 - 2 | 202 | 61 | |
XM_021477452.1 | XP_021333127.1 | 1 - 2 | 558 | 61 | ||
GenBank mRNA | AY424303.1 | - | 1 - 2 | 202 | 61 | |
AY533544.1 | - | 1 - 2 | 84 | 61 | ||
AY714131.1 | - | 2 - 3 | 210 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Danio rerio Probe spans exons |