Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ltv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002316.2 | NP_001002316.2 | 4 - 5 | 588 | 81 | |
XM_021470934.1 | XP_021326609.1 | 4 - 5 | 583 | 81 | ||
GenBank mRNA | AY648712.1 | - | 4 - 5 | 546 | 81 | |
BC085516.1 | - | 4 - 5 | 561 | 81 | ||
EH504800.1 | - | 4 - 5 | 599 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |