Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc48a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002424.2 | NP_001002424.1 | 2 - 3 | 395 | 94 | |
XM_009296974.3 | XP_009295249.1 | 2 - 3 | 332 | 94 | ||
GenBank mRNA | BC076146.1 | - | 2 - 3 | 396 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |