Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc16a9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003552.1 | NP_001003552.1 | 3 - 4 | 572 | 92 | |
GenBank mRNA | BC078337.1 | - | 3 - 4 | 572 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |