Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: upk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040242.1 | NP_001035332.1 | 7 - 8 | 763 | 75 | |
XM_005158400.4 | XP_005158457.1 | 7 - 8 | 792 | 75 | ||
GenBank mRNA | BC115285.1 | - | 7 - 8 | 763 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |