Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr5a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077272.2 | NP_001070740.2 | 5 - 6 | 1340 | 75 | |
NM_214779.2 | NP_999944.2 | 5 - 6 | 1345 | 75 | ||
GenBank mRNA | AF327373.1 | - | 5 - 6 | 1218 | 75 | |
BC124756.1 | - | 5 - 6 | 1311 | 75 | ||
BC163909.1 | - | 5 - 6 | 1308 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |