Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col11a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083844.1 | NP_001077313.1 | 21 - 22 | 2022 | 80 | |
XM_005162813.1 | XP_005162870.1 | 22 - 23 | 2349 | 80 | ||
XM_005162814.1 | XP_005162871.1 | 22 - 23 | 2346 | 80 | ||
XM_005162815.1 | XP_005162872.1 | 21 - 22 | 2262 | 80 | ||
XM_005162818.1 | XP_005162875.1 | 21 - 22 | 2052 | 80 | ||
XM_021470209.1 | XP_021325884.1 | 20 - 21 | 1965 | 80 | ||
XM_021470210.1 | XP_021325885.1 | 15 - 16 | 1129 | 80 | ||
GenBank mRNA | BC133083.1 | - | 21 - 22 | 2022 | 80 | |
GQ485664.1 | - | 22 - 23 | 2243 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |