Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131113.1 | NP_571188.1 | 8 - 9 | 1238 | 67 | |
XM_005162296.4 | XP_005162353.1 | 7 - 8 | 1364 | 67 | ||
XM_017353502.2 | XP_017208991.1 | 8 - 9 | 1444 | 67 | ||
GenBank mRNA | BC162134.1 | - | 8 - 9 | 1080 | 67 | |
BC162146.1 | - | 8 - 9 | 1080 | 67 | ||
X68432.1 | - | 1238 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |