Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205588.3 | NP_991151.3 | 1 - 2 | 193 | 78 | |
XM_021479142.1 | XP_021334817.1 | 2 - 3 | 273 | 78 | ||
GenBank mRNA | AF510720.1 | - | 1 - 2 | 194 | 78 | |
BC094967.1 | - | 1 - 2 | 207 | 78 | ||
BC164351.1 | - | 1 - 2 | 217 | 78 | ||
EH547457.1 | - | 1 - 2 | 231 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |