Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lman2la | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205725.2 | NP_991288.2 | 1 - 2 | 151 | 72 | |
GenBank mRNA | AF506214.1 | - | 1 - 2 | 186 | 72 | |
BC097031.1 | - | 1 - 2 | 151 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |