Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scinlb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213090.2 | NP_998255.2 | 14 - 15 | 1938 | 103 | |
GenBank mRNA | BC066531.1 | - | 14 - 15 | 1903 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |