Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lipf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213404.1 | NP_998569.1 | 4 - 5 | 469 | 139 | |
GenBank mRNA | BC052131.1 | - | 4 - 5 | 469 | 139 | |
BC154786.1 | - | 4 - 5 | 469 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |