Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kctd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213478.2 | NP_998643.1 | 2 - 3 | 279 | 70 | |
GenBank mRNA | BC061457.1 | - | 2 - 3 | 251 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |