Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002666.1 | NP_001002666.2 | 5 - 6 | 599 | 95 | |
XM_021477460.1 | XP_021333135.1 | 5 - 6 | 738 | 95 | ||
GenBank mRNA | BC076553.1 | - | 5 - 6 | 599 | 95 | |
BC163973.1 | - | 5 - 6 | 513 | 95 | ||
HM138690.1 | - | 5 - 6 | 485 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |