Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpa1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007065.1 | NP_001007066.1 | 11 - 12 | 1225 | 111 | |
XM_017358267.2 | XP_017213756.1 | 10 - 11 | 1220 | 111 | ||
GenBank mRNA | AY677196.1 | - | 11 - 12 | 1225 | 111 | |
EU826643.1 | - | 10 - 11 | 1194 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |