Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079679.3 | NP_001073147.1 | 7 - 8 | 698 | 70 | |
XM_005168711.4 | XP_005168768.1 | 7 - 8 | 739 | 70 | ||
XM_005168712.4 | XP_005168769.1 | 7 - 8 | 696 | 70 | ||
XM_005168713.4 | XP_005168770.1 | 7 - 8 | 739 | 70 | ||
GenBank mRNA | BC124539.1 | - | 6 - 7 | 650 | 70 | |
DT063943.1 | - | 7 - 8 | 681 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |