Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxp3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001329567.1 | NP_001316496.1 | 10 - 11 | 1004 | 89 | |
XM_021478298.1 | XP_021333973.1 | 10 - 11 | 1060 | 89 | ||
GenBank mRNA | BC154484.1 | - | 10 - 11 | 1004 | 89 | |
BC163356.1 | - | 9 - 10 | 983 | 89 | ||
FJ906821.1 | - | 9 - 10 | 910 | 89 | ||
FM881778.1 | - | 9 - 10 | 977 | 89 | ||
FM881779.1 | - | 5 - 6 | 554 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |