Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itgb3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082948.1 | NP_001076417.1 | 13 - 14 | 2212 | 76 | |
GenBank mRNA | BC163207.1 | - | 13 - 14 | 2171 | 76 | |
DQ178646.1 | - | 13 - 14 | 2212 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |