Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104649.2 | NP_001098119.2 | 16 - 17 | 1695 | 155 | |
XM_021471707.1 | XP_021327382.1 | 16 - 17 | 1716 | 155 | ||
XM_021471709.1 | XP_021327384.1 | 15 - 16 | 1506 | 155 | ||
XM_021471716.1 | XP_021327391.1 | 17 - 18 | 1805 | 155 | ||
GenBank mRNA | BC163912.1 | - | 16 - 17 | 1717 | 155 | |
EF653429.1 | - | 16 - 17 | 1777 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |