Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: isl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130964.2 | NP_571039.1 | 2 - 3 | 338 | 76 | |
GenBank mRNA | BC067333.1 | - | 2 - 3 | 317 | 76 | |
BC095011.1 | - | 2 - 3 | 338 | 76 | ||
U09404.1 | - | 1 - 2 | 37 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |