Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197934.1 | NP_922916.1 | 7 - 8 | 1387 | 90 | |
GenBank mRNA | AF533661.1 | - | 7 - 8 | 1387 | 90 | |
BC049303.1 | - | 6 - 7 | 1187 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |