Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199591.2 | NP_955885.1 | 10 - 11 | 1614 | 91 | |
XM_005160454.3 | XP_005160511.1 | 11 - 12 | 1738 | 91 | ||
GenBank mRNA | AY577010.1 | - | 11 - 12 | 1681 | 91 | |
BC066614.1 | - | 10 - 11 | 1614 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |