Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gorab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200385.1 | NP_956679.1 | 4 - 5 | 750 | 72 | |
XM_005160649.4 | XP_005160706.1 | 4 - 5 | 800 | 72 | ||
GenBank mRNA | BC053289.1 | - | 4 - 5 | 750 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |