Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacna1da | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203484.1 | NP_982351.1 | 21 - 22 | 2804 | 118 | |
XM_021479576.1 | XP_021335251.1 | 21 - 22 | 2992 | 118 | ||
XM_021479578.1 | XP_021335253.1 | 21 - 22 | 2992 | 118 | ||
XM_021479579.1 | XP_021335254.1 | 21 - 22 | 2992 | 118 | ||
XM_021479581.1 | XP_021335256.1 | 20 - 21 | 2916 | 118 | ||
XM_021479583.1 | XP_021335258.1 | 21 - 22 | 2992 | 118 | ||
XM_021479584.1 | XP_021335259.1 | 20 - 21 | 2916 | 118 | ||
GenBank mRNA | AY528224.1 | - | 21 - 22 | 2804 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |