Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC079485.1 | - | 9 - 10 | 1048 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |