Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030116.1 | NP_001025287.1 | 4 - 5 | 581 | 84 | |
XM_005160433.1 | XP_005160490.1 | 4 - 5 | 637 | 84 | ||
XM_005160434.1 | XP_005160491.1 | 4 - 5 | 637 | 84 | ||
GenBank mRNA | BC124746.1 | - | 4 - 5 | 584 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |