Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mylka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009304445.1 | XP_009302720.1 | 20 - 21 | 3746 | 60 | |
XM_009304446.1 | XP_009302721.1 | 20 - 21 | 3746 | 60 | ||
XM_017357739.1 | XP_017213228.1 | 20 - 21 | 3743 | 60 | ||
GenBank mRNA | AB267908.1 | - | 5 - 6 | 1103 | 60 | |
BC163913.1 | - | 5 - 6 | 891 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |