Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: selenow1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178287.5 | NP_840072.3 | 5 - 6 | 415 | 143 | |
GenBank mRNA | AY216582.1 | - | 5 - 6 | 326 | 143 | |
BC092686.1 | - | 5 - 6 | 377 | 143 | ||
BC152097.1 | - | 5 - 6 | 351 | 143 | ||
BI472564.1 | - | 1 - 2 | 346 | 143 | ||
CK395720.1 | - | 5 - 6 | 426 | 143 | ||
EH445855.1 | - | 5 - 6 | 438 | 143 | ||
EH564833.1 | - | 1 - 2 | 427 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |