Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199578.1 | NP_955872.1 | 6 - 7 | 799 | 76 | |
GenBank mRNA | BC059660.1 | - | 6 - 7 | 799 | 76 | |
BC071334.1 | - | 6 - 7 | 795 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |