Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pak1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200945.2 | NP_957239.2 | 9 - 10 | 1110 | 72 | |
GenBank mRNA | AY391445.1 | - | 9 - 10 | 1110 | 72 | |
BC049528.1 | - | 9 - 10 | 1158 | 72 | ||
BC066416.1 | - | 9 - 10 | 1117 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |