Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mgst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045302.2 | NP_001038767.1 | 2 - 3 | 186 | 70 | |
GenBank mRNA | BC095059.1 | - | 2 - 3 | 286 | 70 | |
BC117619.1 | - | 2 - 3 | 188 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |