Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: extl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076561.1 | NP_001070029.1 | 3 - 4 | 544 | 114 | |
GenBank mRNA | BC124426.1 | - | 3 - 4 | 544 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |